Få mere at vide
Acetate Standard: CH3CO2- @ 10000 μg/mL in H2O, VHG™
Artikelnummer. 17958274
Skift visning
Mængde:
500 ml
Artikelnummer. Mængde
17958274 500 ml
1 options
Artikelnummer. 17958274 Leverandør VHG Labs Leverandørnr. VHGI1PACET500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Acetate Standard: CH3CO2- @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetate
CAS 71-50-1
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnavne Acetate
Forsendelsestilstand Cold 2°c to 8°C
Molekylær formel C2 H3 O2
Synonym (C2 H3 O)2-
InChI formel InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/p-1
SMIL CC(=O)[O-]
IUPAC navn acetate
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 59.044
Mængde 500 ml
Formel vægt 59.013 g/mol