Få mere at vide
Acetarsone, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acetarsone |
| CAS | 97-44-9 |
| Molekylær formel | C8H10AsNO5 |
| Mængde | 5 g |
| Synonym | As-[3-(Acetylamino)-4-hydroxyphenyl]arsonic Acid, 190F, 3-Acetamido-4-hydroxybenzenearsonic Acid, 3-Acetamido-4-hydroxyphenylarsonic Acid, 3-Acetylamino-4-hydroxyphenyl-1-arsonic Acid, 3-Acetylamino-4-hydroxyphenylarsonic Acid, Acetarsol, Acetarsone, Acetphenarsine, Amarsan, Amoebal, Arsaphen, Arsonine, Chrlich 594, Devegan, Disparicida, Dynarsan, Ehrlich 594, F 190, Fourneau 190, Ginarsol, Goyl, Gynoplix, Kharophen, Kubarsol, Limarsol, Malagride, Mexyl, Monargan, N-Acetyl-4-hydroxy-m-arsanilic Acid, NSC 13160, NSC 3247, Nilacid, Oralcid, Orarsan, Osarsal, Osarsol, Osarsole, Osvarsan, Pallicid, Paroxyl, SVC, Sanogyl, Spirocid, Spirozid, Stovarsol, Stovarsolan, Vagisept, Vagoflor |
| InChI formel | InChI=1S/C8H10AsNO5/c1-5(11)10-7-4-6(9(13,14)15)2-3-8(7)12/h2-4,12H,1H3,(H,10,11)(H2,13,14,15) |
| SMIL | CC(=O)Nc1cc(ccc1O)[As](=O)(O)O |
| IUPAC navn | (3-acetamido-4-hydroxyphenyl)arsonic acid |
| Molekylvægt (g/mol) | 275.09 |
| Formel vægt | 274.9775 |