Få mere at vide
Acetanilide-d8, TRC
Artikelnummer. 30310439
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30310439 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310439 Leverandør Toronto Research Chemicals Leverandørnr. TRCA168331500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetanilide-d8
CAS 22778-17-2
Molekylær formel C8 D8 H N O
Mængde 500 mg
Synonym Acetamide-2,2,2-d3, N-(phenyl-2,3,4,5,6-d5)-, Acetamide-2,2,2-d3, N-(phenyl-d5)- (9CI), Acetanilide-2,2,2,2',3',4',5',6'-d8 (8CI), N-(Phenyl-2,3,4,5,6-d5)acetamide-2,2,2-d3, Acetanilide-d8, Acetanilide-D8 (acetyl-D3, phenyl-D5), N-(Acetyl-d3)aniline-2,3,4,5,6-d5, 2,2,2,2',3',4',5',6'-Octadeuteroacetanilide
InChI formel InChI=1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)/i1D3,2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(NC(=O)C([2H])([2H])[2H])c([2H])c1[2H]
IUPAC navn 2,2,2-trideuterio-N-(2,3,4,5,6-pentadeuteriophenyl)acetamide
Molekylvægt (g/mol) 143.213
Formel vægt 143.119