Få mere at vide
Acetanilide-d8, CDN
Artikelnummer. 30314558
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314558 1 g -
30316806 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314558 Leverandør CDN Leverandørnr. CDND51091G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetanilide-d8
CAS 22778-17-2
Molekylær formel C8 D8 H N O
Mængde 1 g
Synonym Acetamide-2,2,2-d3, N-(phenyl-2,3,4,5,6-d5)-, Acetamide-2,2,2-d3, N-(phenyl-d5)- (9 CI), Acetanilide-2,2,2,2',3',4',5',6'-d8 (8 CI), N-(Phenyl-2,3,4,5,6-d5)acetamide-2,2,2-d3, Acetanilide-d8, 2,2,2,2',3',4',5',6'-Octadeuteroacetanilide, 2,2,2,2',3',4',5',6'-Octadeuteroacetanilide, 2,2,2,2',3',4',5',6'-Octadeuteroacetanilide
InChI formel InChI=1 S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6 H,1H3,(H,9,10)/i1D3,2 D,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c([2 H])c(NC(=O)C([2 H])([2 H])[2 H])c([2 H])c1[2 H]
IUPAC navn 2,2,2-trideuterio-N-(2,3,4,5,6-pentadeuteriophenyl)acetamide
Molekylvægt (g/mol) 143.213
Formel vægt 143.119 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity