Få mere at vide
Acetanilide-d6, TRC
Artikelnummer. 30309340
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30309340 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309340 Leverandør Toronto Research Chemicals Leverandørnr. TRCA161352500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetanilide-d6
CAS 141801-46-9
Molekylær formel C8 D6 H3 N O
Mængde 500 mg
Synonym Acetamide-N-d, N-(phenyl-d5)- (9CI), Acetanilide-N,2',3',4',5',6'-d6, N-(Phenyl-2,3,4,5,6-d5)acetamide-N-d1, Acetanilide-d6, Acetanilide D6 (N D1, phenyl-D5), N-Acetylaniline-N,2,3,4,5,6-d6, N,2',3',4',5',6'-Hexadeuteroacetanilide
InChI formel InChI=1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)/i2D,3D,4D,5D,6D/hD
SMIL [2H]N(C(=O)C)c1c([2H])c([2H])c([2H])c([2H])c1[2H]
IUPAC navn N-deuterio-N-(2,3,4,5,6-pentadeuteriophenyl)acetamide
Molekylvægt (g/mol) 141.2
Formel vægt 141.1061