missing translation for 'onlineSavingsMsg'
Få mere at vide

Acetaminotadalafil, TRC

Artikelnummer. 30135457
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135457 25 mg -
30004261 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30135457 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16125025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1446144-71-3
Molekylær formel C23 H20 N4 O5
Molekylvægt (g/mol) 432.43
Synonym Acetamide, N-[(6R,12aR)-6-(1,3-benzodioxol-5-yl)-3,4,6,7,12,12a-hexahydro-1,4-dioxopyrazino[1',2':1,6]pyrido[3,4-b]indol-2(1H)-yl]-, N-[(6R,12aR)-6-(1,3-Benzodioxol-5-yl)-3,4,6,7,12,12a-hexahydro-1,4-dioxopyrazino[1',2':1,6]pyrido[3,4-b]indol-2(1H)-yl]acetamide, Acetaminotadalafil
IUPAC navn N-[(2R,8R)-2-(1,3-benzodioxol-5-yl)-4,7-dioxo-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-6-yl]acetamide
SMIL CC(=O)NN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O

Specifications

Kemisk navn eller materiale Acetaminotadalafil
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C23H20N4O5/c1-12(28)25-26-10-20(29)27-17(23(26)30)9-15-14-4-2-3-5-16(14)24-21(15)22(27)13-6-7-18-19(8-13)32-11-31-18/h2-8,17,22,24H,9-11H2,1H3,(H,25,28)/t17-,22-/m1/s1
Formel vægt 432.1434
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.