Få mere at vide
Acetaminophen Dimer-d6, TRC
Artikelnummer. 30325775
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30325775 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325775 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1612372.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetaminophen Dimer-d6
CAS 1794817-30-3
Molekylær formel C16H10D6N2O4
Mængde 2.5 mg
Synonym N,N'-(6,6'-Dihydroxy[1,1'-biphenyl]-3,3'-diyl)bisacetamide-d6, Paracetamol Dimer Impurity-d6
InChI formel InChI=1S/C16H16N2O4/c1-9(19)17-11-3-5-15(21)13(7-11)14-8-12(18-10(2)20)4-6-16(14)22/h3-8,21-22H,1-2H3,(H,17,19)(H,18,20)/i1D3,2D3
SMIL OC1=C(C2=CC(NC(C([2H])([2H])[2H])=O)=CC=C2O)C=C(NC(C([2H])([2H])[2H])=O)C=C1
IUPAC navn 2,2,2-trideuterio-N-[4-hydroxy-3-[2-hydroxy-5-[(2,2,2-trideuterioacetyl)amino]phenyl]phenyl]acetamide
Molekylvægt (g/mol) 306.35
Formel vægt 306.1487