Få mere at vide
Acetaminophen-d4 (major), TRC
Artikelnummer. 30323805
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30323805 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323805 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16122225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Paracetamol-D4
CAS 64315-36-2
Renhedsgrad noter HPLC
Molekylær formel C8 2H4 H5 N O2
Mængde 25 mg
Synonym Paracetamol D4 (phenyl D4), Paracetamol D4, Paracetamol-d4, Acetaminophen D4, Acetaminophen-d4, Acetamide, N-(4-hydroxyphenyl-2,3,5,6-d4)-, 4-Hydroxy-2,3,5,6-tetradeuteroacetanilide, N-(4-Hydroxyphenyl-2,3,5,6-d4)acetamide
InChI formel InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(NC(=O)C)c([2H])c([2H])c1O
IUPAC navn N-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetamide
Molekylvægt (g/mol) 155.19
Formel vægt 155.0884
Procent renhed >95