Få mere at vide
Acetaminophen-d3, TRC
Artikelnummer. 30312243
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30312243 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312243 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1612212.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetaminophen methyl-d3
CAS 60902-28-5
Renhedsgrad noter HPLC
Molekylær formel C8 2H3 H6 N O2
Mængde 2.5 mg
Synonym Acetaminophen D3 (Methyl D3), Acetamide-2,2,2-d3, N-(4-hydroxyphenyl)-, N-(4-Hydroxyphenyl)acetamide-2,2,2-d3, Paracetamol D3, Paracetamol D3 (Methyl D3)
InChI formel InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)/i1D3
SMIL [2H]C([2H])([2H])C(=O)Nc1ccc(O)cc1
IUPAC navn 2,2,2-trideuterio-N-(4-hydroxyphenyl)acetamide
Molekylvægt (g/mol) 154.18
Formel vægt 154.0822
Procent renhed >95