Få mere at vide
Acetamide-d5, TRC
Artikelnummer. 30324466
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30324466 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324466 Leverandør Toronto Research Chemicals Leverandørnr. TRCA133061100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetamide-d5
CAS 33675-83-1
Molekylær formel C2 D5 N O
Mængde 100 mg
Synonym Acetamide-d5
InChI formel InChI=1S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)/i1D3/hD2
SMIL [2H]N([2H])C(=O)C([2H])([2H])[2H]
IUPAC navn N,N,2,2,2-pentadeuterioacetamide
Molekylvægt (g/mol) 64.098
Formel vægt 64.0685