missing translation for 'onlineSavingsMsg'
Learn More

Acetamide-2,2,2-d3, TRC

Product Code. 30326519
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30326519 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30326519 Supplier Toronto Research Chemicals Supplier No. TRCA18979550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Acetamide-2,2,2-d3
CAS 23724-60-9
Molekylær formel C2 D3 H2 N O
Mængde 50 mg
Synonym 2,2,2-Trideuteroacetamide, 2,2,2-Trideuterioacetamide, Acetamide-2,2,2-d3 (7CI, 8CI, 9CI)
InChI formel InChI=1S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)/i1D3
SMIL [2H]C([2H])([2H])C(=O)N
IUPAC navn 2,2,2-trideuterioacetamide
Molekylvægt (g/mol) 62.09
Formel vægt 62.0559
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.