Få mere at vide
Acetamide-2,2,2-d3, TRC
Artikelnummer. 30315287
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30315287 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315287 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18979510MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetamide-2,2,2-d3
CAS 23724-60-9
Molekylær formel C2 D3 H2 N O
Mængde 10 mg
Synonym 2,2,2-Trideuteroacetamide, 2,2,2-Trideuterioacetamide, Acetamide-2,2,2-d3 (7CI, 8CI, 9CI)
InChI formel InChI=1S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)/i1D3
SMIL [2H]C([2H])([2H])C(=O)N
IUPAC navn 2,2,2-trideuterioacetamide
Molekylvægt (g/mol) 62.09
Formel vægt 62.0559