Få mere at vide
Acetaldehyde Diethyl Acetal-d4, TRC
Artikelnummer. 30318665
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30318665 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318665 Leverandør Toronto Research Chemicals Leverandørnr. TRCA132723100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetaldehyde Diethyl Acetal-d4
CAS 352438-71-2
Molekylær formel C6 D4 H10 O2
Mængde 100 mg
Synonym Acetaldehyde Diethylacetal D4 (1,1,1,2-D4), Ethane-1,1,1,2-d4, 2,2-diethoxy- (9CI)
InChI formel InChI=1S/C6H14O2/c1-4-7-6(3)8-5-2/h6H,4-5H2,1-3H3/i3D3,6D
SMIL [2H]C([2H])([2H])C([2H])(OCC)OCC
IUPAC navn 1,1,1,2-tetradeuterio-2,2-diethoxyethane
Molekylvægt (g/mol) 122.199
Formel vægt 122.124