Få mere at vide
Acetaldehyde-d3, TRC
Artikelnummer. 30311035
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30311035 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311035 Leverandør Toronto Research Chemicals Leverandørnr. TRCA13260125MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetaldehyde-d3
CAS 19901-15-6
Molekylær formel C2 D3 H O
Mængde 25 mg
Synonym Acetic Aldehyde-d3, Ethanal-d3, Ethyl Aldehyde-d3, Perdeuteroacetaldehyde-d3, Tetradeuteroacetaldehyde-d3, Acetaldehyde-2,2,2-d3
InChI formel InChI=1S/C2H4O/c1-2-3/h2H,1H3/i1D3
SMIL [2H]C([2H])([2H])C=O
IUPAC navn 2,2,2-trideuterioacetaldehyde
Molekylvægt (g/mol) 47.07
Formel vægt 47.045