Få mere at vide
Acesulfame-d4 Potassium Salt, TRC
Artikelnummer. 30308891
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30308891 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308891 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1325021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acesulfame-d4 Potassium Salt
CAS 1623054-53-4
Renhedsgrad noter HPLC
Molekylær formel C4D4KNO4S
Mængde 1 mg
Synonym 6-Methyl-1,2,3-oxathiazin-4(3H)-one-d4 2,2-Dioxide Potassium Salt, Acesulfam H-d4 Potassium Salt, 3,4-Dihydro-6-methyl-1,2,3-oxathiazin-4-one-d4 2,2-Dioxide Potassium Salt
InChI formel InChI=1S/C4H5NO4S.K/c1-3-2-4(6)5-10(7,8)9-3;/h2H,1H3,(H,5,6);/q;+1/p-1/i1D3,2D;
SMIL O=S1(OC(C([2H])([2H])[2H])=C([2H])C(N1[K])=O)=O
IUPAC navn potassium;5-deuterio-2,2-dioxo-6-(trideuteriomethyl)-1-oxa-2λ6-thia-3-azanidacyclohex-5-en-4-one
Molekylvægt (g/mol) 205.27
Formel vægt 204.9749
Procent renhed >95