Få mere at vide
Acepromazine-d6 Maleate, TRC
Artikelnummer. 30322083
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30322083 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322083 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1309021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acepromazine D6 maleate
CAS 1331655-50-5
Molekylær formel C19 D6 H16 N2 OS . C4 H4 O4
Mængde 1 mg
Synonym Acepromazine D6 Maleate (N,N-dimethyl D6), 1-[10-[3-(Bis(trideuteriomethyl)amino)propyl]phenothiazin-2-yl]ethanone, but-2-enedioic acid
InChI formel InChI=1S/C19H22N2OS.C4H4O4/c1-14(22)15-9-10-19-17(13-15)21(12-6-11-20(2)3)16-7-4-5-8-18(16)23-19;5-3(6)1-2-4(7)8/h4-5,7-10,13H,6,11-12H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/i2D3,3D3;
SMIL [2H]C([2H])([2H])N(CCCN1c2ccccc2Sc3ccc(cc13)C(=O)C)C([2H])([2H])[2H].OC(=O)\C=C/C(=O)O
IUPAC navn 1-[10-[3-[bis(trideuteriomethyl)amino]propyl]phenothiazin-2-yl]ethanone;(Z)-but-2-enedioic acid
Molekylvægt (g/mol) 448.56
Formel vægt 448.1939