Få mere at vide
Acephate-d6, TRC
Artikelnummer. 30312985
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30312985 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312985 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1308521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acephate-d6
CAS 2109699-73-0
Renhedsgrad noter GC
Molekylær formel C42 H6 H4 N O3 P S
Mængde 1 mg
Synonym Acetylphosphoramidothioic Acid O,S-Dimethyl Ester, Ortho 12420, Orthene
InChI formel InChI=1S/C4H10NO3PS/c1-4(6)5-9(7,8-2)10-3/h1-3H3,(H,5,6,7)/i2D3,3D3
SMIL [2H]C([2H])([2H])OP(=O)(NC(=O)C)SC([2H])([2H])[2H]
IUPAC navn N-[trideuteriomethoxy(trideuteriomethylsulfanyl)phosphoryl]acetamide
Molekylvægt (g/mol) 189.203
Formel vægt 189.0496
Procent renhed >95