Få mere at vide
Acephate-d3 (N-Acetyl-d3), CDN
Artikelnummer. 30310232
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30310232 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310232 Leverandør CDN Leverandørnr. CDND81790.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acephate D3 (acetyl D3)
CAS 2140327-70-2
Molekylær formel C4 D3 H7 N O3 P S
Mængde 0.01 g
Synonym Acephate D3 (acetyl D3), Acephate-d3 (N-Acetyl-d3)
InChI formel InChI=1 S/C4H10NO3PS/c1-4(6)5-9(7,8-2)10-3/h1-3H3,(H,5,6,7)/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)NP(=O)(OC)SC
IUPAC navn 2,2,2-trideuterio-N-[methoxy(methylsulfanyl)phosphoryl]acetamide
Molekylvægt (g/mol) 186.1843
Formel vægt 186.0307 g/mol
Procent renhed 99 atom % D, min 95% Chemical Purity