Få mere at vide
Acenocoumarol-d4, TRC
Artikelnummer. 30326269
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30326269 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326269 Leverandør Toronto Research Chemicals Leverandørnr. TRCA13080210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acenocoumarol-d5
CAS 1185071-64-0
Molekylær formel C19 D4 H11 N O6
Mængde 10 mg
InChI formel InChI=1S/C19H15NO6/c1-11(21)10-15(12-6-8-13(9-7-12)20(24)25)17-18(22)14-4-2-3-5-16(14)26-19(17)23/h2-9,15,22H,10H2,1H3/i6D,7D,8D,9D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1C(CC(=O)C)C2=C(O)c3ccccc3OC2=O)[N+](=O)[O-]
IUPAC navn 4-hydroxy-3-[3-oxo-1-(2,3,5,6-tetradeuterio-4-nitrophenyl)butyl]chromen-2-one
Molekylvægt (g/mol) 357.35
Formel vægt 357.115