Få mere at vide
Acenaphthene-d10, TRC
Artikelnummer. 30376374
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30376374 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376374 Leverandør Toronto Research Chemicals Leverandørnr. TRCD448334100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acenaphthene D10
CAS 15067-26-2
Molekylær formel C12 2H10
Mængde 100 mg
Synonym Acenaphthylene-d8, 1,2-dihydro-d2-, Acenaphthene-d10 (6CI,7CI,8CI), 1,2-Dihydro-d2acenaphthylene-d8, Acenaphthene D10
InChI formel InChI=1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2/i1D,2D,3D,4D,5D,6D,7D2,8D2
SMIL [2H]c1c([2H])c2c3c(c([2H])c([2H])c([2H])c3c1[2H])C([2H])([2H])C2([2H])[2H]
IUPAC navn 1,1,2,2,3,4,5,6,7,8-decadeuterioacenaphthylene
Molekylvægt (g/mol) 164.27
Formel vægt 164.141