Få mere at vide
Acenaphthene-d10, CDN
Artikelnummer. 30312902
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312902 5 g
30319254 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312902 Leverandør CDN Leverandørnr. CDND00425G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acenaphthene D10
CAS 15067-26-2
Molekylær formel C12 2H10
Mængde 5 g
InChI formel InChI=1 S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6 H,7-8H2/i1D,2 D,3 D,4 D,5 D,6 D,7D2,8D2
SMIL [2 H]c1c([2 H])c2c3c(c([2 H])c([2 H])c([2 H])c3c1[2 H])C([2 H])([2 H])C2([2 H])[2 H]
IUPAC navn 1,1,2,2,3,4,5,6,7,8-decadeuterioacenaphthylene
Molekylvægt (g/mol) 164.27
Formel vægt 164.141 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity