Få mere at vide
Acemetacin, Thermo Scientific™
A potent inhibitor of COX-2 found to have anti-tumor activity in the colon
476.13 DKK - 2082.12 DKK
Kemiske identifikatorer
| CAS | 53164-05-9 |
|---|---|
| Molekylær formel | C21H18ClNO6 |
| Molekylvægt (g/mol) | 415.826 |
| MDL nummer | MFCD00151473 |
| InChI nøgle | FSQKKOOTNAMONP-UHFFFAOYSA-N |
| Synonym | acemetacin, rantudil, emflex, acemetacinum, acemix, acemetacina, acemetacine, aximeixin, rheumibis, indomethacin carboxymethyl ester |
| PubChem CID | 1981 |
| ChEBI | CHEBI:31162 |
| IUPAC navn | 2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid |
| SMIL | CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC(=O)O |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15475779
|
Thermo Scientific Alfa Aesar
J63583.04 |
2g |
476.13 DKK
2g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15485779
|
Thermo Scientific Alfa Aesar
J63583.09 |
10g |
2082.12 DKK
10g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| 53164-05-9 | |
| 415.826 | |
| FSQKKOOTNAMONP-UHFFFAOYSA-N | |
| 1981 | |
| 2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid |
| C21H18ClNO6 | |
| MFCD00151473 | |
| acemetacin, rantudil, emflex, acemetacinum, acemix, acemetacina, acemetacine, aximeixin, rheumibis, indomethacin carboxymethyl ester | |
| CHEBI:31162 | |
| CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC(=O)O |
Tekniske data
| Acemetacin | |
| C21H18ClNO6 | |
| UN2811 | |
| acemetacin, rantudil, emflex, acemetacinum, acemix, acemetacina, acemetacine, aximeixin, rheumibis, indomethacin carboxymethyl ester | |
| CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC(=O)O | |
| 415.826 | |
| CHEBI:31162 |
| 53164-05-9 | |
| MFCD00151473 | |
| 14,27 | |
| FSQKKOOTNAMONP-UHFFFAOYSA-N | |
| 2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid | |
| 1981 | |
| 415.83 |
Sikkerhed og håndtering
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 258-403-4
missing translation for 'rtecsNumber' : NL3521400
missing translation for 'tsca' : No
Anbefalet opbevaring : Store at -20°C