Få mere at vide
Acebutolol-d7, TRC
Artikelnummer. 30327449
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30327449 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327449 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1238031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acebutolol-d7
CAS 2701782-36-5
Molekylær formel C18 D7 H21 N2 O4
Mængde 1 mg
Synonym Acebutolol-d7, (±)-Acebutolol-d7 (iso-propyl-d7), Acebutolol-D7 (N-isopropyl-D7), N-[3-Acetyl-4-[2-hydroxy-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]propoxy]phenyl]butanamide
InChI formel InChI=1S/C18H28N2O4/c1-5-6-18(23)20-14-7-8-17(16(9-14)13(4)21)24-11-15(22)10-19-12(2)3/h7-9,12,15,19,22H,5-6,10-11H2,1-4H3,(H,20,23)/i2D3,3D3,12D
SMIL [2H]C([2H])([2H])C([2H])(NCC(O)COc1ccc(NC(=O)CCC)cc1C(=O)C)C([2H])([2H])[2H]
IUPAC navn N-[3-acetyl-4-[3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-2-hydroxypropoxy]phenyl]butanamide
Molekylvægt (g/mol) 343.4691
Formel vægt 343.2488