Få mere at vide
Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns™ Cartridge Columns
Artikelnummer. 11362223 Shop alle Thermo Scientific produkter
Længde (metrisk):
5 mm
15 mm
Pakningsstørrelse:
Pakke med 5
Artikelnummer. 11362223
Brand: Thermo Scientific™ 163592
Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns™ Cartridge Columns are very short microcolumns consisting of a set of disposable cartridges. Cartridge holder sold separately (p/n 164650).
Thermo Scientific™ Acclaim™ 5μm PepMap™ 300 μ-Precolumns™ Cartridge Columns are very short microcolumns consisting of a set of disposable cartridges. Cartridge holder sold separately (p/n 164650).
  • For sample pre-concentration, sample cleanup, or in various column-switching techniques
  • 300Å pore size
TRUSTED_SUSTAINABILITY
Tekniske data
Kolonneformat Pre-Column Traps
Diameter (metrisk) 1 mm
Til brug med LC/MS, ESI-MS, MALDI-MS
Længde (metrisk) 5 mm
Partikelstørrelse 5 μm
pH 2 to 8
Porestørrelse 300 Å
Overfladeareal 100 m²/g
USP type L1
Stationær fase C18
Produktlinje nanoLC
Emballagemateriale Silica
Kulstofbelastning 9%
Kolonnetype Peptide Analysis
Fase Reversed Phase
Materiale Stainless Steel