Få mere at vide
WTW™ Accessories for WTW™ Handheld Multiparameter
:
pH Electrode
Tekniske data
Tekniske data
| Konnektortyper | S7 |
| Knudepunkt | Pin Hole |
| Materiale | POM Armoring, Glass Shaft, Lucalen Watering Cap, PSU S7 Protective Cap |
| pH-område | 4 to 12 |
| Temperatur (metrisk) Drift | 0°C to 50°C |
| Type | Armored pH Electrode |
| Til brug med (applikation) | Measurements in normally loaded surface water and waste water |
| Model- | SensoLyt™ MPP-A |
| Opbevaringskrav | With watering cap, filled with KCl (3mol/L), and protective cap for S7 plug-in connection |
| Temperaturområde (metrisk). | 0°C to 50°C |