Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Cimarec+ and SuperNuova+ Series Hotplates
Add functionality to your Thermo Scientific™ Cimarec+ and SuperNuova+ Series Hotplates with these accessories.
359.00 DKK - 570.00 DKK
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15463660 View Documents Thermo Scientific™
CIC0001447
PT100 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers 359.00 DKK
Each View alternative products
Save Up to 
»
Please contact our Product Support team for availability on +45 70 27 99 20
Add to basket
15493660 View Documents Thermo Scientific™
CIC0001449
Hastelloy PT1000 External probe, 0-400°C, Hastelloy, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers 570.00 DKK
Each View alternative products
Save Up to 
»
Please contact our Product Support team for availability on +45 70 27 99 20
Add to basket
15483660 View Documents Thermo Scientific™
CIC0001448
PT1000 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers 385.00 DKK
Each View alternative products
Save Up to 
»
Please contact our Product Support team for availability on +45 70 27 99 20
Add to basket