Learn More
Cytiva Accessories for SE 660 Vertical Electrophoresis Unit
Brand:  Cytiva SE6102
644.70 DKK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.53800kg
 View more versions of this product
Product Code. 11520764
  • 679.63 DKK / Pack of 2
Estimated Shipment: 21-05-2024
to see stock.
Specifications
SE 660 Vertical Electrophoresis Unit
Glass
18 cm
Glass Plate
2/Pk.
16 cm