Learn More
Thermo Scientific™ Filtration Hardware, Accessories, and Spare Parts
Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in a wide range of applications and in conjunction with our various lines of filtration devices.
Brand: Thermo Scientific™ DS0345-0001
8952.86 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 4.43000kg
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Features- Outlets spaced for easy manipulation of individual filter funnels
- Teflon™ stopcock on each valve shuts off flow when fewer than three outlets are used
- Ideal for use with Nalgene reusable filter funnel with clamp
- For use with Nalgene analytical filter funnels or Nagene test filter funnels
Includes:
Three vacuum outlets, each with two-way valve and vent port; hose barb vacuum connector for 0.38″ (9.5mm) I.D. tubing.
Note:
These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Vacuum Filter Funnel Manifold | |
Stainless Steel | |
Vacuum Manifold |