Learn More
Thermo Scientific™ Filtration Hardware, Accessories, and Spare Parts
Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in a wide range of applications and in conjunction with our various lines of filtration devices.
Brand: Thermo Scientific™ 710300-0006
108.47 DKK valid until 2024-03-30
Use promo code "21648" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Features
- Graduated receiver allows easy visualization of sample volume
- Two side arms allow connection to vacuum line
- Upper chamber is designed for pressure or vacuum filtration
- Independent locking rings seal upper chamber to receiver without damagingmembrane to ensure efficient filtration
- Chamber accommodates 47 or 50mmmembranes (not included) and has three ports for venting (with or without syringe filters) and aseptic addition of samples
- Ports also can be attached to a pressure source
- Upper chamber is supplied with twomembrane support plates: a sterilization plate and an analytical plate
Receiver Accepts
- 0.25 to 0.3125 in. I.D. (6.4 to 8mm) vacuum tubing
- Syringe filters
Sterilization Plate (one-piece, clear)
- Providesmaximum flowrate and throughput
- Suitable for cold sterilization or clarification of culturemedia, reagents or solvents that are compatible with holder andmembranematerials
- Nominal filter area, 13.3cm2
Analytical Plate (two-piece, white)
- Provides optimalmembrane support and keepsmembrane flat to facilitate analytical procedures
- Suitable for analysis of particulates and biological contaminants, or anymicrobiological analyses using themembrane filter technique
- Nominal filter area, 11.3cm2
Includes:
Complete upper chamber, receiver, polypropylene cover and TPE port caps. (membranes not included.)
Specifications
Vacuum Tubing Adapters |