Learn More
BINDER™ Back Access Port (50 mm)
For Use With: BINDER units series ED
Brand:  BINDER™ 8012-0546
Product Code. 15641786
  • 4960.00 DKK / Each
Only null left
Estimated Shipment: 03-09-2025
Description
  • For use in back
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Access port with silicone plug - 50 mm, back
Steel
Silicone plug
BINDER Chambers Series ED
Access Port
50 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Back Access Port (50 mm) > Diameter: 50 mm, For Use With: All ED, FD, FED, FP, M models