missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ACATE2 Recombinant Protein

Brand:  Abnova™ H00023597-P01.25ug

 View more versions of this product

Product Code. 16137522

  • 4145.00 DKK / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH12573
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
49.06
8
Glutathione Sepharose 4 Fast Flow
25 μg
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACOT9
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
23597
ACATE2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
ACATE2/CGI-16/MT-ACT48
ACOT9
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ ACATE2 Recombinant Protein > 25μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.