Learn More
R&D Systems™ Ac4GlcNAlk
Description
Ac4GlcNAlk is a metabolic chemical reporter (MCR) for studying cell surface glycosylation. It is used for identification of O-GlcNAc modified proteins. In cells expressing the engineered pyrophosphorylase, mut-AGX1, Ac4GlcNAlk is efficiently converted into UDP-GlcNAlk which is incorporated into GlcNAlk-containing glycoproteins.
Specifications
Specifications
| Chemical Name or Material | 2-Deoxy-2-[(1-oxo-4-pentyn-1-yl)amino]-D-glucopyranose-1,3,4,6-tetraacetate |
| CAS | 1361993-37-4 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Glycobiology Probes |
| Molecular Formula | C19H25NO10 |
| Purity | 0.95 |