missing translation for 'onlineSavingsMsg'
Få mere at vide

Ac-lys(ac)-OH, TRC

Artikelnummer. 30179195
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179195 1 g
30179047 100 mg
30178980 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30179195 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1692401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-86-5
Molekylær formel C10 H18 N2 O4
Molekylvægt (g/mol) 230.26
Synonym N2,N6-Diacetyl-L-lysine, Diacetyl-L-lysine, Nalpha,Nepsilon-Diacetyl-L-lysine, alpha,epsilon-Diacetyl-L-lysine;
IUPAC navn N2,N6-diacetyl-L-lysine
SMIL [C@@H](CCCCNC(C)=O)(NC(C)=O)C(O)=O

Specifications

Kemisk navn eller materiale Ac-lys(ac)-OH
InChI formel InChI=1S/C10H18N2O4/c1-7(13)11-6-4-3-5-9(10(15)16)12-8(2)14/h9H,3-6H2,1-2H3,(H,11,13)(H,12,14)(H,15,16)/t9-/m0/s1
Mængde 1 g
Formel vægt 230.13
Forsendelsestilstand Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.