Få mere at vide
Ac-lys(ac)-OH, TRC
Artikelnummer. 30179047
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179047 100 mg
30178980 500 mg
30179195 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179047 Leverandør Toronto Research Chemicals Leverandørnr. TRCA169240100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 499-86-5
Molekylær formel C10 H18 N2 O4
Molekylvægt (g/mol) 230.26
Synonym N2,N6-Diacetyl-L-lysine, Diacetyl-L-lysine, Nalpha,Nepsilon-Diacetyl-L-lysine, alpha,epsilon-Diacetyl-L-lysine;
IUPAC navn N2,N6-diacetyl-L-lysine
SMIL [C@@H](CCCCNC(C)=O)(NC(C)=O)C(O)=O
Tekniske data
Kemisk navn eller materiale Ac-lys(ac)-OH
InChI formel InChI=1S/C10H18N2O4/c1-7(13)11-6-4-3-5-9(10(15)16)12-8(2)14/h9H,3-6H2,1-2H3,(H,11,13)(H,12,14)(H,15,16)/t9-/m0/s1
Mængde 100 mg
Formel vægt 230.13