missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Ac-DEVD-AFC

Product Code. 16441998
Encompass_Preferred
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16441998 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441998 Supplier MedChemExpress Supplier No. HYP1005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ac-DEVD-AFC is a fluorogenic substrate (λex=400 nm, λem=530 nm).

Specifications

CAS 201608-14-2
Opløselighed DMSO : ≥ 50 mg/mL (68.53 mM) ∣H2O : < 0.1 mg/mL (insoluble)
Indhold og opbevaring Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Til brug med (applikation) Cancer-programmed cell death
Mængde 5 mg
Molekylvægt (g/mol) 729.61 g/mol
Produkttype Dye Reagents
Renhed eller kvalitetsklasse 99.15%
Forsendelsestilstand Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.