Få mere at vide
Tocris Bioscience™ AC 265347
CaSR biased allosteric modulator
Brand: Tocris Bioscience™ 6165/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
AC 265347 is a CaSR biased allosteric modulator (pEC50 = 7.8-8.1). Calcimimetic. Displays ability to bias signalling towards the accumulation of pERK1/2 and IP1. Reduces parathyroid hormone (PTH) levels in rat serum without inducing the release of calcitonin. Exhibits no significant activation of human GABAB or type I PTH receptors. Orally active.Tekniske data
| α-(2,4-Dimethylphenyl)-α-methyl-2-benzothiazolemethanol | |
| 1253901-26-6 | |
| >99% | |
| 50 mg |