Få mere at vide
Tocris Bioscience™ AC 1903
Beskrivelse
AC 1903 is a selective TRPC5 blocker (IC50 = 13.6 μM). Also displays weak inhibition at TRPC4 (IC50 >100 μM) but not TRPC6. Exhibits no effect against a standard panel of profiling kinases. Blocks TRPC5 channel activity in glomeruli of proteinuric rats. Suppresses severe proteinuria and prevents podocyte loss in a transgenic rat model of focal segmental glomerulosclerosis (FSGS).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(2-Furanylmethyl)-1-(phenylmethyl)-1H-benzimidazol-2-amine |
| CAS | 831234-13-0 |
| Mængde | 25 mg |
| Molekylær formel | C19H17N3O |
| Formel vægt | 303.36 |
| Renhed | 98% |