missing translation for 'onlineSavingsMsg'
Learn More
ABT-751, MedChemExpress Product Code.: 16466146

ABT-751, MedChemExpress

Product Code. p-200099487
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16466146

Brand: Medchem Express HY13270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ABT-751(E 7010) is a novel bioavailable tubulin-binding and antimitotic sulfonamide agent with IC50 of about 1.5 and 3.4 ÎĽM in neuroblastoma and non-neuroblastoma cell lines, respectively.

ABT-751(E 7010) is a novel bioavailable tubulin-binding and antimitotic sulfonamide agent with IC50 of about 1.5 and 3.4 ÎĽM in neuroblastoma and non-neuroblastoma cell lines, respectively.

Specifications

Chemical Name or Material ABT-751
CAS 141430-65-1
Color White
Purity Grade Notes Research
Molecular Formula C18H17N3O4S
Quantity 100 mg
Synonym E7010
Solubility Information DMSO : ≥ 48 mg/mL (129.24 mM)
SMILES COC1=CC=C(C=C1)S(=O)(NC2=C(N=CC=C2)NC3=CC=C(C=C3)O)=O
Molecular Weight (g/mol) 371.41
Formula Weight 371.41
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
ABT-751, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.