Få mere at vide
ABT 263-d8, TRC
Artikelnummer. 30179259
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179259 1 mg
30179305 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179259 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1125021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217620-38-6
Molekylær formel C47H47D8ClF3N5O6S3
Molekylvægt (g/mol) 982.66
Synonym 4-[4-[[2-(4-Chlorophenyl)-5,5-dimethyl-1-cyclohexen-1-yl]methyl]-1-piperazinyl]-N-[[4-[[(1R)-3-(4-morpholinyl)-1-[(phenylthio)methyl]propyl]amino]-3-[(trifluoromethyl)sulfonyl]phenyl]sulfonyl]benzamide-d8;
IUPAC navn 4-[4-[[2-(4-chlorophenyl)-5,5-dimethylcyclohexen-1-yl]methyl]piperazin-1-yl]-N-[4-[[(2R)-4-(2,2,3,3,5,5,6,6-octadeuteriomorpholin-4-yl)-1-phenylsulfanylbutan-2-yl]amino]-3-(trifluoromethylsulfonyl)phenyl]sulfonylbenzamide
SMIL O=C(NS(C1=CC(S(=O)(C(F)(F)F)=O)=C(N[C@@H](CSC2=CC=CC=C2)CCN3C([2H])([2H])C([2H])([2H])OC([2H])([2H])C3([2H])[2H])C=C1)(=O)=O)C(C=C4)=CC=C4N(CC5)CCN5CC6=C(C7=CC=C(Cl)C=C7)CCC(C)(C)C6
Tekniske data
Kemisk navn eller materiale ABT 263-d8
Mængde 1 mg
InChI formel InChI=1S/C47H55ClF3N5O6S3/c1-46(2)20-18-42(34-8-12-37(48)13-9-34)36(31-46)32-55-22-24-56(25-23-55)39-14-10-35(11-15-39)45(57)53-65(60,61)41-16-17-43(44(30-41)64(58,59)47(49,50)51)52-38(19-21-54-26-28-62-29-27-54)33-63-40-6-4-3-5-7-40/h3-17,30,38,52H,18-29,31-33H2,1-2H3,(H,53,57)/t38-/m1/s1/i26D2,27D2,28D2,29D2