Få mere at vide
(+)-Abscisic Aldehyde, TRC
Artikelnummer. 30138146
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138146 10 mg
30008625 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138146 Leverandør Toronto Research Chemicals Leverandørnr. TRCA11000510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 41944-86-9
Molekylær formel C15H20O3
Synonym (2Z,4E)-5-[(1S)-1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl]-3-methyl-2,4-pentadienal∣(+)-Abscisyl aldehyde∣Abscisic Aldehyde
Tekniske data
Kemisk navn eller materiale (+)-Abscisic Aldehyde
Mængde 10 mg
Forsendelsestilstand Room Temperature
Formel vægt 248.1412
Renhed >95% (HPLC)
Molekylvægt (g/mol) 248.32
InChI nøgle RIKWDZWVHUIUAM-KICRZJJPSA-N
SMIL C\C(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C=O
IUPAC navn (2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienal