missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fluorescent In Situ Hybridization (FISH) Buffer

Optimized for performance with FISH probes

Brand:  Abnova™ U0029.5x1mL

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16092368

  • 4235.00 DKK / Pack of 5
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fluorescent In Situ Hybridization (Cell)

Specifications

Specifications

Store at 4°C
Fluorescent In Situ Hybridization (Cell)
5 x 1 mL
FISH Hybridization Buffer is optimized for performance with FISH probes and contains dextran sulfate, formamide, and SSC (pH 7.0).
FISH Hybridization Buffer(5ml)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers