Få mere at vide
Abn-CBD, Tocris Bioscience™
Beskrivelse
Neurobehaviorally inactive cannabinoid that acts as a selective agonist for GPR55 (EC50 values are 2.5, >30 and >30 μM at GPR55, CB1 and CB2 receptors respectively). Increases phosphorylation of protein kinases in, and migration of, human umbilical vein endothelial cells.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Abn-CBD |
| CAS | 22972-55-0 |
| Molekylær formel | C21H30O2 |
| Mængde | 10 mg |
| Synonym | abnormal cannabidiol, abn-cbd, 4-1r,6r-3-methyl-6-prop-1-en-2-yl cyclohex-2-en-1-yl-5-pentylbenzene-1,3-diol, e---4-p-mentha-1,8-dien-3-yl-5-pentylresorcinol, resorcinol, 4-p-mentha-1,8-dien-3-yl-5-pentyl-,--e, abnormal-cannabidiol, d0s3um, 4-1r,6r-3-methyl-6-1-methylethenyl-2-cyclohexen-1-yl-5-pentyl-1,3-benzenediol, 4-1r,6r-3-methyl-6-prop-1-en-2-yl-1-cyclohex-2-enyl-5-pentylbenzene-1,3-diol, 4-1r,6r-3-methyl-6-prop-1-en-2-ylcyclohex-2-en-1-yl-5-pentylbenzene-1,3-diol |
| InChI nøgle | YWEZXUNAYVCODW-RBUKOAKNSA-N |
| SMIL | CCCCCC1=CC(=CC(=C1C2C=C(CCC2C(=C)C)C)O)O |
| IUPAC navn | 4-[(1R,6R)-3-methyl-6-prop-1-en-2-ylcyclohex-2-en-1-yl]-5-pentylbenzene-1,3-diol |
| Molekylvægt (g/mol) | 314.469 |
| PubChem CID | 89949 |
| Vis mere |