missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ ABCG5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578701
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: MCF-7 whole cell, A549 whole cell, HT1080 whole cell, U87 whole cell, SKOV whole cell. Flow: A431 cell.
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the White subfamily. The protein encoded by this gene functions as a half-transporter to limit intestinal absorption and promote biliary excretion of sterols. It is expressed in a tissue-specific manner in the liver, colon, and intestine. This gene is tandemly arrayed on chromosome 2, in a head-to-head orientation with family member ABCG8. Mutations in this gene may contribute to sterol accumulation and atherosclerosis, and have been observed in patients with sitosterolemia.
Tekniske data
| ABCG5 | |
| Polyclonal | |
| Unconjugated | |
| ABCG5 | |
| Abcg5; ATP binding cassette subfamily G member 5; ATP-binding cassette sub-family G (WHITE) member 5 (sterolin 1); ATP-binding cassette sub-family G member 5; ATP-binding cassette, subfamily G (WHITE), member 5; ATP-binding cassette, sub-family G (WHITE), member 5; ATP-binding cassette, sub-family G (WHITE), member 5 (sterolin 1); ATP-binding cassette, subfamily G, member 5; AW112016; cmp; sterolin 1; sterolin-1; STSL; trac | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64240 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| Q9H222 | |
| ABCG5 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human ABCG5 (116-131aa NGRALRREQFQDCFSY). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion