Få mere at vide
Novus Biologicals™ ABCB5 Antibody (Brigham and Women's patent anti-ABCB5) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28518-1mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ABCB5 Monoclonal antibody specifically detects ABCB5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| ABCB5 | |
| Monoclonal | |
| Unconjugated | |
| Q2M3G0 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Brigham and Women's patent anti-ABCB5 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
| ABCB5 | |
| 1 mg | |
| ABC Transporters, Lipid and Metabolism | |
| 340273 | |
| Human | |
| Purified |