Learn More
Abcam Rat monoclonal [23G3] Anti-Mouse IgE epsilon chain (Biotin), 250 ug
Product Code. 32109544 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
Product Code. Quantity Unit Size
32109544 250 μg 250µg
1 options
Product Code. 32109544 Supplier Abcam Supplier No. ab99585250ug
 Restricted Item
Forventet forsendelse: 02-12-2026
Rat Monoclonal antibody, suitable for ICC/IF, Flow Cyt, ELISA.
  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: ICC/IF, Flow Cyt, ELISA
  • Species Reactivity: Mouse
  • Storage Buffer: pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 23G3
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
Gene IGHE
Gene Symbols IGHE
Host Species Rat
Purification Method Affinity purification
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule IGHE, Mouse
Validated Application abrv ICC/IF, Flow Cyt, ELISA