Learn More
Abcam Rat IL-1Ra Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32114960 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32114960 10 x 96 Tests 96 tests
1 options
Product Code. 32114960 Supplier Abcam Supplier No. ab25373310x96Tests
 Restricted Item
Forventet forsendelse: 26-11-2026
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Rat
Especificaciones
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols Il1rn
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Rt
Validated Application abrv sELISA
Mostrar más Mostrar menos