Learn More
Abcam Rabbit F(ab')2 Anti-Cow IgG H&L (HRP), 500 uL
Product Code. 32111182 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
Product Code. Quantity Unit Size
32111182 500 μL 500µL
1 options
Product Code. 32111182 Supplier Abcam Supplier No. ab136757500uL
 Restricted Item
Forventet forsendelse: 27-11-2026
Rabbit F(ab')2 Anti-Cow IgG H&L (HRP)
  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ELISA, Flow Cyt, ICC, IHC-Fr, IHC-P, WB
  • Species Reactivity: Bovine
  • Storage Buffer: Constituents: 50% Glycerol (glycerin, glycerine), 50% Tris buffered saline
  • Form: Liquid
  • Purification: Affinity purification Immunogen
Specifications
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation Constituents: 50% Glycerol (glycerin, glycerine), 50% Tris buffered saline
Host Species Rabbit
Purification Method Affinity purification Immunogen
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Cow.
Target Species Bovine
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Cow
Validated Application abrv Flow Cyt, IHC-Fr, ICC, IHC-P, WB, ELISA