Learn More
Abcam Rabbit Anti-Mouse IgG H&L (Alkaline Phosphatase) preadsorbed, 500 ug
Product Code. 32108829 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Product Code. Quantity Unit Size
32108829 500 μg 500µg
1 options
Product Code. 32108829 Supplier Abcam Supplier No. ab7077500ug
 Restricted Item
Forventet forsendelse: 27-11-2026
Rabbit Anti-Mouse IgG H&L (Alkaline Phosphatase) preadsorbed
  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Applications: Dot, ELISA, ICC/IF, IHC-Fr, IHC-P, IM, WB
  • Species Reactivity: Mouse
  • Storage Buffer: BSA, Glycerol, Sodium chloride, Tris HCl, Magnesium chloride, Zinc chloride
  • Form: Liquid
  • Purification: Affinity purification
Specifications
Applications Dot Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.01% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 1% BSA, 0.87% Sodium chloride, 0.79% Tris HCl, 0.0095% Magnesium chloride, 0.00136% Zinc chloride
Gene Symbols Ighg1
Host Species Rabbit
Purification Method Affinity purification
Quantity 500 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Polyclonal Secondary Antibody
Form Liquid
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv Dot, IHC-Fr, IHC-P, IM, ICC/IF, ELISA, WB