Learn More
Abcam Rabbit Anti-Human IgG2 H&L (Biotin), 150 ug
Product Code. 32117255 Shop All Abcam Products
Change view
Click to view available options
Quantity:
150 μg
Unit Size:
150µg
Product Code. Quantity Unit Size
32117255 150 μg 150µg
1 options
Product Code. 32117255 Supplier Abcam Supplier No. ab86254150ug
 Restricted Item
Forventet forsendelse: 27-11-2026
Rabbit Polyclonal secondary antibody, suitable for WB, ELISA, RIA.
  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: WB, ELISA, RIA
  • Species Reactivity: Human
  • Storage Buffer: pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine), 0.25% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein G
Specifications
Applications Western Blot, ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Biotin
Formulation pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine), 0.25% BSA
Gene Symbols IGHG2
Host Species Rabbit
Purification Method Affinity purification Protein G
Quantity 150 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv WB, ELISA, RIA