Learn More
Abcam Mouse Transferrin Receptor Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32112685 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32112685 10 x 96 Tests 96 tests
1 options
Product Code. 32112685 Supplier Abcam Supplier No. ab25663110x96Tests
 Restricted Item
Forventet forsendelse: 26-11-2026
Antibody, suitable for sELISA, with 1 citations.
  • Applications: sELISA
  • Species Reactivity: Mouse
Specifications
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols Tfrc
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv sELISA